Assay Details
| Gene Symbol: mc5rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173280.1 | NP_775387.1 | 1 | 652 | 76 | |
| XM_005158169.4 | XP_005158226.1 | 2 | 789 | 76 | ||
| GenBank mRNA | BC163224.1 | - | 1 | 76 | ||
| BC163233.1 | - | 1 | 76 | |||
More Information
Set Membership: |
|

mc5rb
Genomic Map