Assay Details
| Gene Symbol: chrnb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173287.1 | NP_775394.1 | 2 - 3 | 209 | 68 | |
| GenBank mRNA | AB087185.1 | - | 2 - 3 | 209 | 68 | |
| BC163409.1 | - | 2 - 3 | 278 | 68 | ||
| BC163411.1 | - | 2 - 3 | 278 | 68 | ||
More Information
Set Membership: |
|

chrnb3b
Genomic Map