Assay Details
| Gene Symbol: selenow1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178287.5 | NP_840072.3 | 4 - 5 | 312 | 106 | |
| GenBank mRNA | AY216582.1 | - | 4 - 5 | 223 | 106 | |
| BC092686.1 | - | 4 - 5 | 274 | 106 | ||
| BC152097.1 | - | 4 - 5 | 248 | 106 | ||
| BI472564.1 | - | 2 - 3 | 449 | 106 | ||
| CK395720.1 | - | 4 - 5 | 323 | 106 | ||
| EH445855.1 | - | 4 - 5 | 335 | 106 | ||
| EH564833.1 | - | 2 - 3 | 530 | 106 | ||
More Information
Set Membership: |
|

selenow1
Genomic Map