Assay Details
| Gene Symbol: krt18a.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178437.2 | NP_848524.1 | 2 - 3 | 596 | 70 | |
| GenBank mRNA | AF537368.1 | - | 2 - 3 | 578 | 70 | |
| AJ493269.1 | - | 2 - 3 | 300 | 70 | ||
| BC045869.1 | - | 2 - 3 | 596 | 70 | ||
| BC065848.1 | - | 2 - 3 | 543 | 70 | ||
| BC164730.1 | - | 2 - 3 | 550 | 70 | ||
More Information
Set Membership: |
|

krt18a.1
Genomic Map