Assay Details
| Gene Symbol: krt18a.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178437.2 | NP_848524.1 | 4 - 5 | 912 | 71 | |
| GenBank mRNA | AF537368.1 | - | 4 - 5 | 894 | 71 | |
| AJ493269.1 | - | 4 - 5 | 616 | 71 | ||
| BC045869.1 | - | 4 - 5 | 912 | 71 | ||
| BC065848.1 | - | 4 - 5 | 859 | 71 | ||
| BC164730.1 | - | 4 - 5 | 866 | 71 | ||
More Information
Set Membership: |
|

krt18a.1
Genomic Map