Assay Details
| Gene Symbol: tmpoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328563.1 | NP_001315492.1 | 13 - 14 | 1648 | 76 | |
| NM_180968.3 | NP_851299.1 | 13 - 14 | 1564 | 76 | ||
| XM_005164721.4 | XP_005164778.1 | 10 - 11 | 1270 | 76 | ||
| GenBank mRNA | AJ320189.1 | - | 13 - 14 | 1597 | 76 | |
| AJ320190.1 | - | 13 - 14 | 1512 | 76 | ||
| BC048056.1 | - | 13 - 14 | 1564 | 76 | ||
| BC165860.1 | - | 13 - 14 | 1540 | 76 | ||
More Information
Set Membership: |
|

tmpoa
Genomic Map