Assay Details
| Gene Symbol: tmpoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328563.1 | NP_001315492.1 | 3 - 4 | 561 | 95 | |
| NM_001328565.1 | NP_001315494.1 | 3 - 4 | 561 | 95 | ||
| NM_180968.3 | NP_851299.1 | 3 - 4 | 561 | 95 | ||
| XM_005164720.4 | XP_005164777.1 | 3 - 4 | 568 | 95 | ||
| XM_005164721.4 | XP_005164778.1 | 3 - 4 | 567 | 95 | ||
| XM_005164722.4 | XP_005164779.1 | 3 - 4 | 567 | 95 | ||
| GenBank mRNA | AJ320189.1 | - | 3 - 4 | 509 | 95 | |
| AJ320190.1 | - | 3 - 4 | 509 | 95 | ||
| AJ320191.1 | - | 3 - 4 | 509 | 95 | ||
| AJ320192.1 | - | 3 - 4 | 507 | 95 | ||
| BC048056.1 | - | 3 - 4 | 561 | 95 | ||
| BC165860.1 | - | 3 - 4 | 537 | 95 | ||
More Information
Set Membership: |
|

tmpoa
Genomic Map