Assay Details
| Gene Symbol: tmpoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328563.1 | NP_001315492.1 | 4 - 5 | 692 | 146 | |
| NM_180968.3 | NP_851299.1 | 4 - 5 | 692 | 146 | ||
| XM_005164720.4 | XP_005164777.1 | 4 - 5 | 699 | 146 | ||
| GenBank mRNA | AJ320189.1 | - | 4 - 5 | 640 | 146 | |
| AJ320190.1 | - | 4 - 5 | 640 | 146 | ||
| BC048056.1 | - | 4 - 5 | 692 | 146 | ||
| BC165860.1 | - | 4 - 5 | 668 | 146 | ||
More Information
Set Membership: |
|

tmpoa
Genomic Map