Assay Details
| Gene Symbol: tmpoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328563.1 | NP_001315492.1 | 6 - 7 | 867 | 80 | |
| NM_180968.3 | NP_851299.1 | 6 - 7 | 867 | 80 | ||
| XM_005164720.4 | XP_005164777.1 | 6 - 7 | 874 | 80 | ||
| GenBank mRNA | AJ320190.1 | - | 6 - 7 | 815 | 80 | |
| BC048056.1 | - | 6 - 7 | 867 | 80 | ||
| BC165860.1 | - | 6 - 7 | 843 | 80 | ||
More Information
Set Membership: |
|

tmpoa
Genomic Map