Assay Details
| Gene Symbol: ret | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181662.2 | NP_858048.2 | 19 - 20 | 3267 | 60 | |
| GenBank mRNA | AF007949.1 | - | 19 - 20 | 3269 | 60 | |
| BC163588.1 | - | 19 - 20 | 3264 | 60 | ||
More Information
Set Membership: |
|

ret
Genomic Map