Assay Details
| Gene Symbol: eif4a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198366.2 | NP_938180.1 | 4 - 5 | 385 | 68 | |
| GenBank mRNA | BC048899.1 | - | 4 - 5 | 354 | 68 | |
| EB766529.1 | - | 4 - 5 | 387 | 68 | ||
More Information
Set Membership: |
|

eif4a1a
Genomic Map