Assay Details
| Gene Symbol: LOC108179061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017357051.2 | XP_017212540.1 | 1 - 2 | 326 | 83 | |
| XM_017357052.2 | XP_017212541.1 | 1 - 2 | 326 | 83 | ||
| Gene Symbol: sinhcafl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC055670.1 | - | 1 - 2 | 227 | 83 | |
More Information
Set Membership: |
|

LOC108179061
Genomic Map