Assay Details
| Gene Symbol: aco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198908.1 | NP_944590.1 | 6 - 7 | 842 | 69 | |
| GenBank mRNA | BC058049.1 | - | 6 - 7 | 842 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aco2| Gene Symbol: aco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198908.1 | NP_944590.1 | 6 - 7 | 842 | 69 | |
| GenBank mRNA | BC058049.1 | - | 6 - 7 | 842 | 69 | |
Set Membership: |
|