Assay Details
| Gene Symbol: adam17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199673.1 | NP_955967.1 | 13 - 14 | 1914 | 105 | |
| XM_005158718.4 | XP_005158775.1 | 13 - 14 | 2074 | 105 | ||
| GenBank mRNA | BC057410.1 | - | 13 - 14 | 1914 | 105 | |
More Information
Set Membership: |
|

adam17a
Genomic Map