Assay Details
| Gene Symbol: bzw1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199708.1 | NP_956002.1 | 5 - 6 | 505 | 92 | |
| GenBank mRNA | BC058875.1 | - | 5 - 6 | 505 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bzw1a| Gene Symbol: bzw1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199708.1 | NP_956002.1 | 5 - 6 | 505 | 92 | |
| GenBank mRNA | BC058875.1 | - | 5 - 6 | 505 | 92 | |
Set Membership: |
|