Assay Details
| Gene Symbol: fbxo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199718.2 | NP_956012.1 | 4 - 5 | 440 | 64 | |
| XM_005156487.4 | XP_005156544.1 | 4 - 5 | 441 | 64 | ||
| GenBank mRNA | BC063957.1 | - | 4 - 5 | 387 | 64 | |
| EH446633.1 | - | 4 - 5 | 438 | 64 | ||
More Information
Set Membership: |
|

fbxo9
Genomic Map