Assay Details
| Gene Symbol: atf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199723.1 | NP_956017.1 | 5 - 6 | 611 | 72 | |
| GenBank mRNA | BC053252.1 | - | 5 - 6 | 611 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

atf1| Gene Symbol: atf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199723.1 | NP_956017.1 | 5 - 6 | 611 | 72 | |
| GenBank mRNA | BC053252.1 | - | 5 - 6 | 611 | 72 | |
Set Membership: |
|