Assay Details
| Gene Symbol: lrrc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199747.2 | NP_956041.2 | 1 - 2 | 627 | 75 | |
| GenBank mRNA | BC049443.1 | - | 1 - 2 | 503 | 75 | |
| BC062861.1 | - | 1 - 2 | 582 | 75 | ||
More Information
Set Membership: |
|

lrrc42
Genomic Map