Assay Details
| Gene Symbol: ampd3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199848.1 | NP_956142.1 | 14 - 15 | 2146 | 64 | |
| XM_005159053.4 | XP_005159110.1 | 14 - 15 | 2243 | 64 | ||
| XM_005159054.2 | XP_005159111.1 | 13 - 14 | 2228 | 64 | ||
| XM_005159055.4 | XP_005159112.1 | 13 - 14 | 2147 | 64 | ||
| XM_021467594.1 | XP_021323269.1 | 14 - 15 | 2225 | 64 | ||
| XM_021467595.1 | XP_021323270.1 | 14 - 15 | 2243 | 64 | ||
| GenBank mRNA | BC044154.1 | - | 14 - 15 | 2146 | 64 | |
More Information
Set Membership: |
|

ampd3b
Genomic Map