Assay Details
| Gene Symbol: ampd3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199848.1 | NP_956142.1 | 10 - 11 | 1558 | 83 | |
| XM_005159053.4 | XP_005159110.1 | 10 - 11 | 1655 | 83 | ||
| XM_005159054.2 | XP_005159111.1 | 9 - 10 | 1640 | 83 | ||
| XM_005159055.4 | XP_005159112.1 | 9 - 10 | 1559 | 83 | ||
| XM_021467594.1 | XP_021323269.1 | 10 - 11 | 1637 | 83 | ||
| XM_021467595.1 | XP_021323270.1 | 10 - 11 | 1655 | 83 | ||
| GenBank mRNA | BC044154.1 | - | 10 - 11 | 1558 | 83 | |
More Information
Set Membership: |
|

ampd3b
Genomic Map