Assay Details
| Gene Symbol: capzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199935.1 | NP_956229.1 | 6 - 7 | 805 | 80 | |
| XM_009295745.2 | XP_009294020.1 | 6 - 7 | 756 | 80 | ||
| XM_009295746.3 | XP_009294021.1 | 6 - 7 | 754 | 80 | ||
| GenBank mRNA | BC055514.1 | - | 6 - 7 | 805 | 80 | |
More Information
Set Membership: |
|

capzb
Genomic Map