Assay Details
| Gene Symbol: cox7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199995.1 | NP_956289.1 | 2 - 3 | 278 | 102 | |
| GenBank mRNA | BC059638.1 | - | 2 - 3 | 278 | 102 | |
More Information
Set Membership: |
|

cox7c
Genomic Map