Assay Details
| Gene Symbol: tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328573.1 | NP_001315502.1 | 5 - 6 | 700 | 85 | |
| NM_200015.2 | NP_956309.2 | 5 - 6 | 703 | 85 | ||
| GenBank mRNA | BC047167.1 | - | 5 - 6 | 643 | 85 | |
| BC065630.1 | - | 5 - 6 | 632 | 85 | ||
More Information
Set Membership: |
|

tfg
Genomic Map