Assay Details
Gene Symbol: washc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200183.1 | NP_956477.1 | 6 - 7 | 1381 | 76 | |
GenBank mRNA | BC045490.1 | - | 6 - 7 | 1381 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
