Assay Details
| Gene Symbol: imp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291357.1 | NP_001278286.1 | 2 - 3 | 345 | 66 | |
| GenBank mRNA | EH466588.1 | - | 5 - 6 | 560 | 66 | |
| EH493453.1 | - | 2 - 3 | 410 | 66 | ||
| EH598772.1 | - | 5 - 6 | 591 | 66 | ||
More Information
Set Membership: |
|

imp3
Genomic Map