Assay Details
| Gene Symbol: stx3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200543.1 | NP_956837.1 | 2 - 3 | 112 | 84 | |
| XM_005173107.4 | XP_005173164.1 | 2 - 3 | 492 | 84 | ||
| XM_005173108.4 | XP_005173165.1 | 2 - 3 | 494 | 84 | ||
| XM_005173110.3 | XP_005173167.2 | 2 - 3 | 496 | 84 | ||
| XM_021480990.1 | XP_021336665.1 | 2 - 3 | 494 | 84 | ||
| XM_021480991.1 | XP_021336666.1 | 2 - 3 | 494 | 84 | ||
| XM_021480992.1 | XP_021336667.1 | 2 - 3 | 492 | 84 | ||
| XM_021480993.1 | XP_021336668.1 | 2 - 3 | 493 | 84 | ||
| GenBank mRNA | BC056333.1 | - | 2 - 3 | 112 | 84 | |
More Information
Set Membership: |
|

stx3a
Genomic Map