Assay Details
| Gene Symbol: stx3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200543.1 | NP_956837.1 | 5 - 6 | 358 | 90 | |
| XM_005173107.4 | XP_005173164.1 | 5 - 6 | 738 | 90 | ||
| XM_005173108.4 | XP_005173165.1 | 5 - 6 | 740 | 90 | ||
| XM_005173110.3 | XP_005173167.2 | 5 - 6 | 742 | 90 | ||
| XM_021480990.1 | XP_021336665.1 | 5 - 6 | 740 | 90 | ||
| XM_021480991.1 | XP_021336666.1 | 5 - 6 | 740 | 90 | ||
| XM_021480992.1 | XP_021336667.1 | 5 - 6 | 738 | 90 | ||
| XM_021480993.1 | XP_021336668.1 | 5 - 6 | 739 | 90 | ||
| GenBank mRNA | BC056333.1 | - | 5 - 6 | 358 | 90 | |
| GU189280.1 | - | 3 - 4 | 189 | 90 | ||
More Information
Set Membership: |
|

stx3a
Genomic Map