Assay Details
| Gene Symbol: stx3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200543.1 | NP_956837.1 | 10 - 11 | 876 | 90 | |
| GenBank mRNA | BC056333.1 | - | 10 - 11 | 876 | 90 | |
More Information
Set Membership: |
|

stx3a
Genomic Map