Assay Details
| Gene Symbol: srsf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200593.2 | NP_956887.2 | 1 - 2 | 372 | 64 | |
| GenBank mRNA | BC056752.1 | - | 1 - 2 | 384 | 64 | |
| BC066682.1 | - | 1 - 2 | 372 | 64 | ||
| BC095586.1 | - | 1 - 2 | 389 | 64 | ||
| BC165510.1 | - | 1 - 2 | 223 | 64 | ||
More Information
Set Membership: |
|

srsf1b
Genomic Map