Assay Details
| Gene Symbol: srsf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200593.2 | NP_956887.2 | 3 - 4 | 728 | 79 | |
| GenBank mRNA | BC066682.1 | - | 3 - 4 | 728 | 79 | |
| BC095586.1 | - | 3 - 4 | 745 | 79 | ||
| BC165510.1 | - | 3 - 4 | 579 | 79 | ||
More Information
Set Membership: |
|

srsf1b
Genomic Map