Assay Details
| Gene Symbol: tbl1xr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001326540.1 | NP_001313469.1 | 10 - 11 | 1288 | 120 | |
| GenBank mRNA | BC056790.1 | - | 10 - 11 | 1246 | 120 | |
More Information
Set Membership: |
|

tbl1xr1b
Genomic Map