Assay Details
| Gene Symbol: tbl1xr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001326540.1 | NP_001313469.1 | 7 - 8 | 1070 | 92 | |
| GenBank mRNA | BC056790.1 | - | 7 - 8 | 1028 | 92 | |
More Information
Set Membership: |
|

tbl1xr1b
Genomic Map