Assay Details
| Gene Symbol: edf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200745.1 | NP_957039.1 | 2 - 3 | 225 | 107 | |
| GenBank mRNA | BC059541.1 | - | 2 - 3 | 225 | 107 | |
| BC071480.1 | - | 2 - 3 | 240 | 107 | ||
| BC159201.1 | - | 2 - 3 | 213 | 107 | ||
More Information
Set Membership: |
|

edf1
Genomic Map