Assay Details
| Gene Symbol: rps7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200752.1 | NP_957046.1 | 3 - 4 | 310 | 60 | |
| GenBank mRNA | AY561508.1 | - | 4 - 5 | 362 | 60 | |
| BC059562.1 | - | 3 - 4 | 310 | 60 | ||
More Information
Set Membership: |
|

rps7
Genomic Map