Assay Details
| Gene Symbol: rps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200845.1 | NP_957139.1 | 1 - 2 | 159 | 68 | |
| GenBank mRNA | BC062279.1 | - | 1 - 2 | 159 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rps17| Gene Symbol: rps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200845.1 | NP_957139.1 | 1 - 2 | 159 | 68 | |
| GenBank mRNA | BC062279.1 | - | 1 - 2 | 159 | 68 | |
Set Membership: |
|