Assay Details
| Gene Symbol: pabpc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200882.1 | NP_957176.1 | 13 - 14 | 2278 | 88 | |
| XM_021468020.1 | XP_021323695.1 | 12 - 13 | 2195 | 88 | ||
| GenBank mRNA | BC059662.1 | - | 13 - 14 | 2144 | 88 | |
| BC063948.1 | - | 13 - 14 | 2278 | 88 | ||
More Information
Set Membership: |
|

pabpc1b
Genomic Map