Assay Details
| Gene Symbol: pabpc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200882.1 | NP_957176.1 | 3 - 4 | 965 | 98 | |
| GenBank mRNA | BC059662.1 | - | 3 - 4 | 831 | 98 | |
| BC063948.1 | - | 3 - 4 | 965 | 98 | ||
More Information
Set Membership: |
|

pabpc1b
Genomic Map