Assay Details
| Gene Symbol: pabpc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200882.1 | NP_957176.1 | 7 - 8 | 1432 | 71 | |
| XM_021468020.1 | XP_021323695.1 | 6 - 7 | 1349 | 71 | ||
| GenBank mRNA | BC059662.1 | - | 7 - 8 | 1298 | 71 | |
| BC063948.1 | - | 7 - 8 | 1432 | 71 | ||
More Information
Set Membership: |
|

pabpc1b
Genomic Map