Assay Details
| Gene Symbol: tpd52l2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200885.2 | NP_957179.1 | 3 - 4 | 510 | 74 | |
| XM_005162035.2 | XP_005162092.1 | 3 - 4 | 525 | 74 | ||
| XM_005162036.3 | XP_005162093.1 | 3 - 4 | 525 | 74 | ||
| XM_005162037.2 | XP_005162094.1 | 3 - 4 | 525 | 74 | ||
| XM_021469745.1 | XP_021325420.1 | 3 - 4 | 525 | 74 | ||
| GenBank mRNA | BC063969.1 | - | 3 - 4 | 476 | 74 | |
More Information
Set Membership: |
|

tpd52l2b
Genomic Map