Assay Details
| Gene Symbol: hhatla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200887.1 | NP_957181.1 | 6 - 7 | 835 | 91 | |
| GenBank mRNA | BC063975.1 | - | 6 - 7 | 835 | 91 | |
More Information
Set Membership: |
|

hhatla
Genomic Map