Assay Details
| Gene Symbol: sulf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200936.1 | NP_957230.1 | 12 - 13 | 1930 | 115 | |
| XM_005169269.4 | XP_005169326.1 | 12 - 13 | 2099 | 115 | ||
| XM_005169270.4 | XP_005169327.1 | 12 - 13 | 2229 | 115 | ||
| XM_021479565.1 | XP_021335240.1 | 12 - 13 | 2091 | 115 | ||
| XM_021479566.1 | XP_021335241.1 | 12 - 13 | 2141 | 115 | ||
| GenBank mRNA | BC045403.1 | - | 12 - 13 | 1930 | 115 | |
| BC165233.1 | - | 11 - 12 | 1867 | 115 | ||
More Information
Set Membership: |
|

sulf2a
Genomic Map