Assay Details
| Gene Symbol: sulf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200936.1 | NP_957230.1 | 7 - 8 | 1169 | 67 | |
| XM_005169269.4 | XP_005169326.1 | 7 - 8 | 1338 | 67 | ||
| XM_005169270.4 | XP_005169327.1 | 7 - 8 | 1468 | 67 | ||
| XM_021479565.1 | XP_021335240.1 | 7 - 8 | 1330 | 67 | ||
| XM_021479566.1 | XP_021335241.1 | 7 - 8 | 1380 | 67 | ||
| GenBank mRNA | BC045403.1 | - | 7 - 8 | 1169 | 67 | |
| BC165233.1 | - | 6 - 7 | 1106 | 67 | ||
More Information
Set Membership: |
|

sulf2a
Genomic Map