Assay Details
| Gene Symbol: ptdss1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200956.1 | NP_957250.1 | 6 - 7 | 899 | 77 | |
| GenBank mRNA | BC044533.1 | - | 6 - 7 | 899 | 77 | |
More Information
Set Membership: |
|

ptdss1a
Genomic Map