Assay Details
| Gene Symbol: cd2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200961.1 | NP_957255.1 | 2 - 3 | 135 | 127 | |
| GenBank mRNA | BC045960.1 | - | 2 - 3 | 135 | 127 | |
| BC164209.1 | - | 1 - 2 | 111 | 127 | ||
More Information
Set Membership: |
|

cd2bp2
Genomic Map