Assay Details
| Gene Symbol: qars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201213.1 | NP_957507.1 | 3 - 4 | 542 | 67 | |
| GenBank mRNA | AY648767.1 | - | 3 - 4 | 542 | 67 | |
| BC049420.1 | - | 3 - 4 | 542 | 67 | ||
More Information
Set Membership: |
|

qars1
Genomic Map