Assay Details
| Gene Symbol: zgc:73226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205570.2 | NP_991133.2 | 1 - 2 | 100 | 114 | |
| GenBank mRNA | AF493989.1 | - | 1 - 2 | 121 | 114 | |
| BC059568.1 | - | 1 - 2 | 100 | 114 | ||
More Information
Set Membership: |
|

zgc:73226
Genomic Map