Assay Details
| Gene Symbol: evlb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205633.1 | NP_991196.1 | 4 - 5 | 677 | 86 | |
| XM_021468384.1 | XP_021324059.1 | 4 - 5 | 1130 | 86 | ||
| GenBank mRNA | BC065327.1 | - | 4 - 5 | 677 | 86 | |
| BC155188.1 | - | 4 - 5 | 518 | 86 | ||
More Information
Set Membership: |
|

evlb
Genomic Map