Assay Details
| Gene Symbol: asf1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207063.1 | NP_996946.1 | 3 - 4 | 653 | 64 | |
| GenBank mRNA | BC066417.1 | - | 3 - 4 | 653 | 64 | |
More Information
Set Membership: |
|

asf1ba
Genomic Map