Assay Details
| Gene Symbol: slc1a3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212640.1 | NP_997805.1 | 6 - 7 | 947 | 95 | |
| XM_009301170.2 | XP_009299445.1 | 6 - 7 | 994 | 95 | ||
| GenBank mRNA | BC049340.1 | - | 6 - 7 | 944 | 95 | |
| BC063233.1 | - | 6 - 7 | 947 | 95 | ||
| HM138693.1 | - | 5 - 6 | 843 | 95 | ||
More Information
Set Membership: |
|

slc1a3a
Genomic Map