Assay Details
| Gene Symbol: slc1a3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212640.1 | NP_997805.1 | 8 - 9 | 1385 | 70 | |
| XM_009301170.2 | XP_009299445.1 | 8 - 9 | 1432 | 70 | ||
| GenBank mRNA | BC049340.1 | - | 8 - 9 | 1382 | 70 | |
| BC063233.1 | - | 8 - 9 | 1385 | 70 | ||
| HM138693.1 | - | 7 - 8 | 1281 | 70 | ||
More Information
Set Membership: |
|

slc1a3a
Genomic Map